Gene Transcripts
- Gene Symbol
- Abhd12
- Entrez Gene ID
- 76192
- Gene Name
- abhydrolase domain containing 12
- Gene Aliases
- 1500011G07Rik, 6330583M11Rik, AI431047, AW547313
- Chromosome Location
- Chr. 2: 150832493 - 150904745 on Build GRCm38
- UniGene ID
- Mm.112632
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024465.3 |
NP_077785.2
|
11, 12 |
1206 |
|
GenBank mRNA |
AK160126.1 |
- |
11, 12 |
1210 |
|
AK178612.1 |
- |
4, 5 |
276 |
|
AK185693.1 |
- |
4, 5 |
254 |
|
AK198485.1 |
- |
4, 5 |
276 |
|
AK202769.1 |
- |
4, 5 |
276 |
|
AK206881.1 |
- |
3, 4 |
224 |
|
AK216141.1 |
- |
3, 4 |
193 |
|
AK217770.1 |
- |
4, 5 |
276 |
|
AK219355.1 |
- |
4, 5 |
278 |
|
BC002138.1 |
- |
11, 12 |
1175 |
|
BC002263.1 |
- |
7, 8 |
509 |
3495118 |
View Full Product Details