Gene Transcripts

Gene Symbol
Stard3nl
Entrez Gene ID
76205
Gene Name
STARD3 N-terminal like
Gene Aliases
0610035N01Rik, 6530409L22Rik, AW124774, MENTHO
Chromosome Location
Chr. 13: 19357676 - 19395809 on Build GRCm38
UniGene ID
Mm.41940
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK002760.1 - 5 553
AK018331.1 - 6 554
AK030365.1 - 3 3252
AK152116.1 - 6 616
AK152378.1 - 6 567
BG084215.2 - 6 572

View Full Product Details