Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_080289.1 | NP_525028.1 | 7, 8 | 783 | |
GenBank mRNA | AK004307.1 | - | 7, 8 | 799 | |
AK018293.1 | - | 7, 8 | 797 | ||
AK168351.1 | - | 7, 8 | 780 | ||
AK172201.1 | - | 7, 8 | 751 | ||
AY113690.1 | - | 7, 8 | 757 | ||
BC010194.1 | - | 7, 8 | 783 | 3966541 |