Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029600.3 | NP_083876.3 | 24 | 3502 | |
GenBank mRNA | AF534127.1 | - | 24 | 3505 | |
AF534128.1 | - | 24 | 3430 | ||
AF534129.1 | - | 23 | 3583 | ||
AK149551.1 | - | 24 | 3502 | ||
AK155664.1 | - | 24 | 3480 | ||
AK156615.1 | - | 24 | 3732 | ||
AK172420.1 | - | 24 | 3732 | ||
AY795569.1 | - | 24 | 3502 | ||
AY841885.1 | - | 24 | 3483 | ||
BC046560.1 | - | 21 | 3009 | ||
BC048825.1 | - | 24 | 3434 | ||
BC150788.1 | - | 24 | 3496 | 9087699 |