Gene Transcripts

Gene Symbol
Unc5cl
Entrez Gene ID
76589
Gene Name
unc-5 homolog C (C. elegans)-like
Gene Aliases
2510009H09Rik, BB155270, Zud
Chromosome Location
Chr. 17: 48453839 - 48539709 on Build GRCm38
UniGene ID
Mm.311629
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152823.4 NP_690036.4 6 1208
GenBank mRNA AK144925.1 - 6 1209
AY510108.1 - 5 1075
BC095959.1 - 6 1186 5349225

View Full Product Details