Gene Transcripts
- Gene Symbol
- Dtl
- Entrez Gene ID
- 76843
- Gene Name
- denticleless homolog (Drosophila)
- Gene Aliases
- 2810047L02Rik, 5730564G15Rik, Ramp
- Chromosome Location
- Chr. 1: 191537365 - 191575534 on Build GRCm38
- UniGene ID
- Mm.189102
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB095735.1 |
- |
11, 12 |
1143 |
|
AB095736.1 |
- |
11, 12 |
1143 |
|
AK054412.1 |
- |
11, 12 |
1133 |
|
AK133177.1 |
- |
11, 12 |
1142 |
|
AK154594.1 |
- |
12, 13 |
1183 |
|
AK161401.1 |
- |
8, 9 |
744 |
|
AK164249.1 |
- |
11, 12 |
1143 |
|
AK166351.1 |
- |
11, 12 |
1151 |
|
AK169601.1 |
- |
11, 12 |
1142 |
|
BC060208.1 |
- |
11, 12 |
1102 |
|
View Full Product Details