Gene Transcripts

Gene Symbol
Catsper3
Entrez Gene ID
76856
Gene Name
cation channel, sperm associated 3
Gene Aliases
4921522D01Rik, CACRC
Chromosome Location
Chr. 13: 55784579 - 55824942 on Build GRCm38
UniGene ID
Mm.159795
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252487.1 NP_001239416.1 7 1038
NM_001252488.1 NP_001239417.1 7 885
NM_029772.4 NP_084048.1 7 999
GenBank mRNA AK014942.1 - 7 1000
AY263400.1 - 7 1013
BC089518.1 - 1138 6774298
HM179540.1 - 7 955
HM179541.1 - 7 802

View Full Product Details