Gene Transcripts
- Gene Symbol
- Mnd1
- Entrez Gene ID
- 76915
- Gene Name
- meiotic nuclear divisions 1 homolog (S. cerevisiae)
- Gene Aliases
- 2610034E18Rik, AI591601, Gaj
- Chromosome Location
- Chr. 3: 84087933 - 84156861 on Build GRCm38
- UniGene ID
- Mm.274829
- Species
- Mus musculus
- Gene Symbol
- Mnd1-ps
- Entrez Gene ID
- 100042415
- Gene Name
- Mnd1 retrotransposed pseudogene
- Gene Aliases
- Gm3833
- Chromosome Location
- Chr. 14: 9884095 - 9886868 on Build GRCm38
- UniGene ID
- Mm.235938
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK156325.1 |
- |
1 |
2303 |
|
View Full Product Details