Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_029806.1 | NP_084082.1 | 5 | 622 | |
GenBank mRNA | AK008654.1 | - | 5 | 625 | |
BC107186.2 | - | 5 | 622 | 40050796 | |
BC107187.2 | - | 5 | 622 | 40050800 | |
BC119485.1 | - | 5 | 622 | 40050803 | |
BC119486.1 | - | 5 | 622 | 40050804 |