Gene Transcripts

Gene Symbol
Stk31
Entrez Gene ID
77485
Gene Name
serine threonine kinase 31
Gene Aliases
C330007K24Rik
Chromosome Location
Chr. 6: 49395483 - 49469506 on Build GRCm38
UniGene ID
Mm.128054
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029916.2 NP_084192.2 21 2589
GenBank mRNA AF285580.1 - 21 2589
AK021186.1 - 7 727
BC137989.1 - 21 2586 8861008

View Full Product Details