Gene Transcripts

Gene Symbol
Dcst1
Entrez Gene ID
77772
Gene Name
DC-STAMP domain containing 1
Gene Aliases
A330106H01Rik
Chromosome Location
Chr. 3: 89350235 - 89365239 on Build GRCm38
UniGene ID
Mm.164013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK030269.1 - 8 1115
BC125478.1 - 8 1100 40129993
BC125480.1 - 8 1100 40129995

View Full Product Details