Gene Transcripts

Gene Symbol
Grifin
Entrez Gene ID
77998
Gene Name
galectin-related inter-fiber protein
Gene Aliases
E130101O17Rik
Chromosome Location
Chr. 5: 140563117 - 140567628 on Build GRCm38
UniGene ID
Mm.160040
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030022.1 NP_084298.1 2, 3 104
GenBank mRNA AK021365.1 - 2, 3 105
BC107323.2 - 2, 3 104 40053455
BC107324.1 - 2, 3 104 40053456

View Full Product Details