Gene Transcripts

Gene Symbol
Spink8
Entrez Gene ID
78709
Gene Name
serine peptidase inhibitor, Kazal type 8
Gene Aliases
C630041L24Rik
Chromosome Location
Chr. 9: 109812203 - 109826628 on Build GRCm38
UniGene ID
Mm.99613
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_183136.2 NP_898959.2 3 340
GenBank mRNA AK021276.1 - 3 323
BC048588.1 - 3 340 6707477
CN843460.1 - 3 286
DQ437329.1 - 3 322

View Full Product Details