Gene Transcripts

Gene Symbol
Ndor1
Entrez Gene ID
78797
Gene Name
NADPH dependent diflavin oxidoreductase 1
Gene Aliases
4930447P04Rik, NR1, Ndor
Chromosome Location
Chr. 2: 25244813 - 25255414 on Build GRCm38
UniGene ID
Mm.40020
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001082476.2 NP_001075945.1 6 749
NM_001252541.1 NP_001239470.1 4 548
NM_001252542.1 NP_001239471.1 6 816
GenBank mRNA AK019617.1 - 6 746
AK143124.1 - 6 790
AK163207.1 - 4 518
AK169885.1 - 6 808
BC049789.1 - 6 799 6331033

View Full Product Details