Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023910.6 | NP_076399.4 | 2 | 1083 | |
NM_029805.1 | NP_084081.1 | 2 | 1083 | ||
GenBank mRNA | AF201286.1 | - | 2 | 796 | |
AK018735.2 | - | 2 | 1056 | ||
AK040534.1 | - | 2 | 1020 | ||
AK079510.1 | - | 2 | 809 | ||
AK080828.1 | - | 2 | 1098 | ||
AK143608.1 | - | 2 | 860 | ||
AK144806.1 | - | 1 | 518 | ||
AK150043.1 | - | 2 | 1052 | ||
AK154153.1 | - | 2 | 1033 | ||
AK154490.1 | - | 2 | 683 | ||
AK155712.1 | - | 2 | 1085 | ||
BC018544.1 | - | 1 | 555 | 3666221 | |
BC023761.1 | - | 2 | 987 | 5343927 | |
BC145721.1 | - | 2 | 1046 | 40131024 | |
BC145723.1 | - | 2 | 1046 | 40131026 |