Gene Transcripts

Gene Symbol
Lysmd3
Entrez Gene ID
80289
Gene Name
LysM, putative peptidoglycan-binding, domain containing 3
Gene Aliases
1110030H10Rik, BC003322
Chromosome Location
Chr. 13: 81657657 - 81672871 on Build GRCm38
UniGene ID
Mm.259969
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030257.1 NP_084533.1 3 463
GenBank mRNA BC003322.1 - 3 463

View Full Product Details