Gene Transcripts
- Gene Symbol
- Tnxb
- Entrez Gene ID
- 81877
- Gene Name
- tenascin XB
- Gene Aliases
- Tn-mhc, Tnx
- Chromosome Location
- Chr. 17: 34659788 - 34795460 on Build GRCm38
- UniGene ID
- Mm.290527
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_031176.2 |
NP_112453.2
|
15 |
5701 |
|
GenBank mRNA |
AB235155.1 |
- |
15 |
5701 |
|
View Full Product Details