Gene Transcripts

Gene Symbol
Sp6
Entrez Gene ID
83395
Gene Name
trans-acting transcription factor 6
Gene Aliases
1110025J03Rik, AA591031, AI592962, Epfn, Klf14
Chromosome Location
Chr. 11: 97013569 - 97024739 on Build GRCm38
UniGene ID
Mm.156282
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031183.2 NP_112460.1 2 1779
GenBank mRNA AJ275988.1 - 1 1009
AK137686.1 - 2 2002
AM295492.1 - 2 2023
AY338955.1 - 2 1780
BC138818.1 - 1 1726 8861840
BC138822.1 - 1 1726 8861844

View Full Product Details