Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 19 | 3069 | |
NR_134949.1 | 20 | 3171 | |||
NR_134950.1 | 19 | 3069 | |||
GenBank mRNA | BC140782.1 | - | 19 | 3024 | |
BC144540.1 | - | 20 | 3132 | ||
BC144541.1 | - | 19 | 3024 | ||
D63395.1 | - | 2 | 206 | ||
U95299.1 | - | 19 | 3020 |