Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004557.3 | NP_004548.3 | 12 | 2051 | |
NR_134949.1 | 13 | 2153 | |||
NR_134950.1 | 12 | 2051 | |||
GenBank mRNA | BC140782.1 | - | 12 | 2006 | |
BC144540.1 | - | 13 | 2114 | ||
BC144541.1 | - | 12 | 2006 | ||
U95299.1 | - | 12 | 2002 |