Gene Transcripts

Gene Symbol
Cml2
Entrez Gene ID
93673
Gene Name
camello-like 2
Gene Aliases
Chromosome Location
Chr. 6: 85865422 - 85869137 on Build GRCm38
UniGene ID
Mm.24251
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053096.3 NP_444326.2 3 567
GenBank mRNA AF163315.1 - 3 545
AK050449.1 - 3 497
BC039773.1 - 3 587 4193874

View Full Product Details