Gene Transcripts

Gene Symbol
Amn
Entrez Gene ID
93835
Gene Name
amnionless
Gene Aliases
5033428N14Rik, AV002116
Chromosome Location
Chr. 12: 111271111 - 111276426 on Build GRCm38
UniGene ID
Mm.197639
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033603.2 NP_291081.2 4 295
GenBank mRNA AF320619.1 - 4 295
AK002318.1 - 4 294
AK017204.1 - 4 309
BC087954.1 - 4 308 30119116

View Full Product Details