Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001242378.1 | NP_001229307.1 | 20 | 2798 | |
NM_001242379.1 | NP_001229308.1 | 19 | 2708 | ||
NM_001242380.1 | NP_001229309.1 | 20 | 2798 | ||
NM_001242381.1 | NP_001229310.1 | 19 | 2705 | ||
NM_001242382.1 | NP_001229311.1 | 20 | 2795 | ||
NM_001242383.1 | NP_001229312.1 | 19 | 2705 | ||
NM_033552.3 | NP_291030.2 | 19 | 2708 | ||
GenBank mRNA | AB033759.1 | - | 19 | 2629 | |
AK043017.1 | - | 19 | 2684 | ||
AK220501.1 | - | 20 | 2763 | ||
BC039226.1 | - | 19 | 2657 | 4480841 | |
HM149278.1 | - | 19 | 2669 | ||
HQ533147.1 | - | 20 | 2735 | ||
HQ825316.1 | - | 20 | 2738 | ||
HQ831517.1 | - | 19 | 2645 | ||
JF500487.1 | - | 19 | 2785 | ||
JF500488.1 | - | 20 | 2875 | ||
JF500489.1 | - | 19 | 2785 | ||
JF500490.1 | - | 20 | 2875 | ||
JX220977.1 | - | 20 | 2762 | ||
JX254917.1 | - | 19 | 2675 | ||
JX268544.1 | - | 20 | 2876 |