Gene Transcripts

Gene Symbol
Slc24a3
Entrez Gene ID
94249
Gene Name
solute carrier family 24 (sodium/potassium/calcium exchanger), member 3
Gene Aliases
NCKX3
Chromosome Location
Chr. 2: 145167712 - 145642166 on Build GRCm38
UniGene ID
Mm.217171
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053195.2 NP_444425.1 10 729
GenBank mRNA AF314821.1 - 10 769
BC017615.1 - 10 729 4914291

View Full Product Details