Gene Transcripts
- Gene Symbol
- Lrrc59
- Entrez Gene ID
- 98238
- Gene Name
- leucine rich repeat containing 59
- Gene Aliases
- AA959742, C78668
- Chromosome Location
- Chr. 11: 94629824 - 94645216 on Build GRCm38
- UniGene ID
- Mm.172720
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_133807.1 |
NP_598568.1
|
4, 5 |
611 |
|
GenBank mRNA |
AK053450.1 |
- |
4, 5 |
638 |
|
AK159374.1 |
- |
4, 5 |
667 |
|
AK159552.1 |
- |
4, 5 |
661 |
|
AK166576.1 |
- |
4, 5 |
660 |
|
AK167604.1 |
- |
4, 5 |
602 |
|
AK169390.1 |
- |
4, 5 |
610 |
|
AK186256.1 |
- |
2, 3 |
184 |
|
AK196953.1 |
- |
|
184 |
|
BC006877.1 |
- |
4, 5 |
611 |
2647513 |
BC066172.1 |
- |
4, 5 |
605 |
30429192 |
View Full Product Details