Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173865.3 | NP_776290.1 | 2 | 1223 | |
GenBank mRNA | AK089802.1 | - | 2 | 1224 | |
AK154819.1 | - | 2 | 1196 | ||
BC096596.1 | - | 2 | 1259 | 4239523 | |
BC116280.1 | - | 2 | 1031 | 40094408 | |
BC116281.1 | - | 2 | 1031 | 40094411 |