Gene Transcripts

Gene Symbol
Khdc1b
Entrez Gene ID
98582
Gene Name
KH domain containing 1B
Gene Aliases
AU014713, AU015226, AU024076, AY238603, C85331, C86372, C88118, Khdc1, Khdc1c
Chromosome Location
Chr. 1: 21383556 - 21386384 on Build GRCm38
UniGene ID
Mm.326336
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001113187.1 NP_001106658.1 3 748
GenBank mRNA BG081161.2 - 3 474

View Full Product Details