Gene Transcripts

Gene Symbol
ROR1
Entrez Gene ID
4919
Gene Name
receptor tyrosine kinase like orphan receptor 1
Gene Aliases
NTRKR1, dJ537F10.1
Chromosome Location
Chr. 1: 63774019 - 64181498 on Build GRCh38
UniGene ID
Hs.128753
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083592.1 NP_001077061.1 6 1106
NM_005012.3 NP_005003.2 6 1106
XM_011541526.1 XP_011539828.1 6 705
XM_017001376.1 XP_016856865.1 5 1006
XM_017001377.1 XP_016856866.1 8 3649
GenBank mRNA AK308088.1 - 6 1112
AK309189.1 - 3 348
BC080541.1 - 6 1106
BC128386.1 - 7 716
M97675.1 - 6 1082

View Full Product Details