Gene Transcripts

Gene Symbol
Numa1
Entrez Gene ID
101706
Gene Name
nuclear mitotic apparatus protein 1
Gene Aliases
6720401E04Rik, AA764025, AL022610, AU014979
Chromosome Location
Chr. 7: 101934002 - 102014959 on Build GRCm38
UniGene ID
Mm.27259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133947.3 NP_598708.3 14 2150
GenBank mRNA AK032693.1 - 14 2199
AK050670.1 - 14 2232
AK168399.1 - 14 2168
BC049791.1 - 14 2150 6505037

View Full Product Details