Gene Transcripts

Gene Symbol
Plekhg4
Entrez Gene ID
102075
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 4
Gene Aliases
4931414L13Rik, AI448583, mFLJ00068
Chromosome Location
Chr. 8: 105369441 - 105382862 on Build GRCm38
UniGene ID
Mm.32034
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081333.1 NP_001074802.1 15, 16 2733
GenBank mRNA AK029859.1 - 13, 14 3405

View Full Product Details