Gene Transcripts

Gene Symbol
Cog4
Entrez Gene ID
102339
Gene Name
component of oligomeric golgi complex 4
Gene Aliases
AW554810, D8Ertd515e
Chromosome Location
Chr. 8: 110846675 - 110882234 on Build GRCm38
UniGene ID
Mm.272930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133973.2 NP_598734.1 7 902
GenBank mRNA AK156466.1 - 7 893
AK172465.1 - 7 893
AY223866.1 - 7 929
BC023120.1 - 7 879 4161858

View Full Product Details