Gene Transcripts

Gene Symbol
Slc35e4
Entrez Gene ID
103710
Gene Name
solute carrier family 35, member E4
Gene Aliases
A330108F03Rik, AI414849, AI536442
Chromosome Location
Chr. 11: 3907022 - 3914664 on Build GRCm38
UniGene ID
Mm.171514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153142.3 NP_694782.1 1 1910
GenBank mRNA AK039786.1 - 1 1911
BC020181.1 - 1 944 4488513

View Full Product Details