Gene Transcripts

Gene Symbol
Slc16a6
Entrez Gene ID
104681
Gene Name
solute carrier family 16 (monocarboxylic acid transporters), member 6
Gene Aliases
AW743111, ESTM12, MCT 6, MCT 7, MCT6
Chromosome Location
Chr. 11: 109450855 - 109473596 on Build GRCm38
UniGene ID
Mm.265874
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029842.1 NP_001025013.1 5 1437
NM_134038.2 NP_598799.1 5 1380
GenBank mRNA AK032026.1 - 5 1440
AK170472.1 - 5 1303
BC017129.1 - 5 1371 4506255

View Full Product Details