Gene Transcripts

Gene Symbol
SIX6
Entrez Gene ID
4990
Gene Name
SIX homeobox 6
Gene Aliases
MCOPCT2, ODRMD, OPTX2, Six9
Chromosome Location
Chr. 14: 60509220 - 60511807 on Build GRCh38
UniGene ID
Hs.194756
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007374.2 NP_031400.2 1 629
GenBank mRNA AJ011785.1 - 1 524
BC065831.1 - 1 629
BC069413.1 - 1 533 7262276
BC087852.1 - 1 607

View Full Product Details