Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133352.2 | NP_579930.1 | 12 | 1561 | |
GenBank mRNA | AF269151.1 | - | 12 | 1493 | |
AK075807.1 | - | 12 | 1527 | ||
AK147380.1 | - | 12 | 1556 | ||
AK147629.1 | - | 12 | 1555 | ||
AK220279.1 | - | 12 | 1537 | ||
BC004799.1 | - | 11 | 1190 |