Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002556.2 | NP_002547.1 | 11 | 2304 | |
GenBank mRNA | AF185696.1 | - | 11 | 2304 | |
AK299071.1 | - | 7 | 748 | ||
AK310538.1 | - | 5 | 562 | ||
BC011581.1 | - | 11 | 1892 | 4560111 | |
BC063121.1 | - | 5 | 663 | ||
M86917.1 | - | 11 | 2304 |