Gene Transcripts

Gene Symbol
Galnt2
Entrez Gene ID
108148
Gene Name
UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 2
Gene Aliases
AI480629
Chromosome Location
Chr. 8: 124231394 - 124345722 on Build GRCm38
UniGene ID
Mm.33808
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139272.2 NP_644678.2 10 1064
GenBank mRNA AF348968.1 - 10 932
AK046567.1 - 10 1022
AK145122.1 - 10 942
AK149609.1 - 10 992
AK151536.1 - 10 855
BC007172.1 - 9 799
BC053063.1 - 9 907 5702572
BC059818.1 - 10 1009 5697789

View Full Product Details