Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199274.1 | NP_001186203.1 | 6, 7 | 1170 | |
NM_134017.2 | NP_598778.1 | 6, 7 | 974 | ||
GenBank mRNA | AB070267.1 | - | 6, 7 | 941 | |
AK075600.1 | - | 6, 7 | 875 | ||
AK075947.1 | - | 6, 7 | 909 | ||
AK153126.1 | - | 6, 7 | 928 | ||
AK162087.1 | - | 6, 7 | 909 | ||
BC003457.1 | - | 6, 7 | 905 | 2648452 |