Gene Transcripts

Gene Symbol
Ppp1r15b
Entrez Gene ID
108954
Gene Name
protein phosphatase 1, regulatory (inhibitor) subunit 15b
Gene Aliases
1810033K10Rik, AI606441, C530022L24Rik, CReP
Chromosome Location
Chr. 1: 133131166 - 133139800 on Build GRCm38
UniGene ID
Mm.293628
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133819.3 NP_598580.1 1 1060
GenBank mRNA AK049028.1 - 1 764
AK082957.1 - 1 896
AK086606.1 - 1 692
AK144678.1 - 1 813
AK146089.1 - 1 798
AK167565.1 - 1 883
AK170862.1 - 1 890
BC058078.1 - 1 843

View Full Product Details