Gene Transcripts

Gene Symbol
Rnf20
Entrez Gene ID
109331
Gene Name
ring finger protein 20
Gene Aliases
4833430L21Rik, AW540162, C79397, mKIAA4116
Chromosome Location
Chr. 4: 49632011 - 49656886 on Build GRCm38
UniGene ID
Mm.24765
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163263.1 NP_001156735.1 17 2872
NM_182999.2 NP_892044.1 18 2719
GenBank mRNA AK220492.1 - 17 2788
BC052482.1 - 18 2658 6308280
BQ442966.1 - 5 634
CN531555.1 - 2 118

View Full Product Details