Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026241.3 | NP_080517.1 | 4 | 504 | |
GenBank mRNA | AK018681.1 | - | 4 | 505 | |
AK030874.1 | - | 700 | |||
AK042896.1 | - | 4 | 505 | ||
AK081207.1 | - | 4 | 697 | ||
AK158215.1 | - | 4 | 699 | ||
AK160231.1 | - | 4 | 520 | ||
BC117004.1 | - | 4 | 534 | 40126323 | |
BC119100.1 | - | 4 | 534 | 8733849 | |
BC144893.1 | - | 4 | 569 |