Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289730.1 | NP_001276659.1 | 2 | 307 | |
NM_001289731.1 | NP_001276660.1 | 5 | 863 | ||
NM_133197.4 | NP_573460.2 | 2 | 307 | ||
GenBank mRNA | AB052945.1 | - | 2 | 250 | |
AB101616.1 | - | 2 | 250 | ||
AK032122.1 | - | 2 | 308 | ||
AK045424.1 | - | 5 | 865 | ||
BC125435.1 | - | 2 | 209 | 40129950 |