Gene Transcripts

Gene Symbol
Slc35b1
Entrez Gene ID
110172
Gene Name
solute carrier family 35, member B1
Gene Aliases
Ugalt2
Chromosome Location
Chr. 11: 95384891 - 95391776 on Build GRCm38
UniGene ID
Mm.4593
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016752.1 NP_058032.1 3, 4 428
GenBank mRNA AK002961.1 - 3, 4 422
AK007711.1 - 3, 4 434
AK160124.1 - 3, 4 494
AK183623.1 - 2, 3 212
AK184790.1 - 2, 3 212
AK185566.1 - 2, 3 209
BC002029.1 - 3, 4 409 3592141
D87990.1 - 3, 4 428

View Full Product Details