Gene Transcripts
- Gene Symbol
- MGAT4B
- Entrez Gene ID
- 11282
- Gene Name
- mannosyl (alpha-1,3-)-glycoprotein beta-1,4-N-acetylglucosaminyltransferase, isozyme B
- Gene Aliases
- GNT-IV, GNT-IVB
- Chromosome Location
- Chr. 5: 179797597 - 179806952 on Build GRCh38
- UniGene ID
- Hs.567419
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014275.4 |
NP_055090.1
|
14, 15 |
1996 |
|
NM_054013.3 |
NP_463459.1
|
13, 14 |
2436 |
|
GenBank mRNA |
AB000624.1 |
- |
14, 15 |
1669 |
|
AK023137.1 |
- |
13, 14 |
2436 |
|
AK225490.1 |
- |
11, 12 |
1231 |
|
AK225492.1 |
- |
11, 12 |
1231 |
|
AL162067.1 |
- |
11, 12 |
1099 |
|
AY358984.1 |
- |
14, 15 |
1677 |
|
BC009464.1 |
- |
12, 13 |
1323 |
|
BC051835.1 |
- |
14, 15 |
1872 |
6196058 |
View Full Product Details