Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 179148338-179148839 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3CA
Entrez Gene ID
5290
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit alpha
Gene Aliases
CLOVE, CWS5, MCAP, MCM, MCMTC, PI3K, PI3K-alpha, p110-alpha
UniGene
Hs.553498
Gene Chromosome Location
Chr.3: 179148114-179240093 on Build GRCh38
Transcripts
NM_006218.3 XM_006713658.3

SNP Detail

SNP ID Chromosome Location
rs1045780121 Chr.3: 179148374-179148374 on Build GRCh38
rs970445077 Chr.3: 179148772-179148772 on Build GRCh38
rs964037508 Chr.3: 179148496-179148496 on Build GRCh38
rs2699887 Chr.3: 179148620-179148620 on Build GRCh38
rs976434651 Chr.3: 179148439-179148439 on Build GRCh38
rs902525309 Chr.3: 179148724-179148724 on Build GRCh38
rs902061149 Chr.3: 179148733-179148733 on Build GRCh38
rs1012080080 Chr.3: 179148763-179148763 on Build GRCh38
rs957226537 Chr.3: 179148780-179148780 on Build GRCh38
rs540317705 Chr.3: 179148771-179148771 on Build GRCh38
rs968911829 Chr.3: 179148760-179148760 on Build GRCh38
rs933432070 Chr.3: 179148714-179148714 on Build GRCh38
rs1005039832 Chr.3: 179148422-179148422 on Build GRCh38
rs865911786 Chr.3: 179148599-179148599 on Build GRCh38
rs558523131 Chr.3: 179148546-179148546 on Build GRCh38
rs900585787 Chr.3: 179148690-179148690 on Build GRCh38
rs1020210145 Chr.3: 179148787-179148787 on Build GRCh38
rs200523412 Chr.3: 179148596-179148596 on Build GRCh38
rs869312615 Chr.3: 179148799-179148799 on Build GRCh38
rs981873678 Chr.3: 179148784-179148784 on Build GRCh38
rs888050513 Chr.3: 179148532-179148532 on Build GRCh38
rs553084547 Chr.3: 179148454-179148454 on Build GRCh38
rs913077120 Chr.3: 179148641-179148641 on Build GRCh38
rs572916698 Chr.3: 179148535-179148535 on Build GRCh38
rs183959683 Chr.3: 179148796-179148796 on Build GRCh38
rs528651622 Chr.3: 179148800-179148800 on Build GRCh38
rs139255255 Chr.3: 179148539-179148539 on Build GRCh38
rs935859667 Chr.3: 179148457-179148457 on Build GRCh38
rs1039216559 Chr.3: 179148655-179148655 on Build GRCh38
rs202171889 Chr.3: 179148519-179148519 on Build GRCh38
rs879484472 Chr.3: 179148393-179148393 on Build GRCh38
rs567363722 Chr.3: 179148419-179148419 on Build GRCh38
rs199579153 Chr.3: 179148585-179148585 on Build GRCh38
rs1049807529 Chr.3: 179148532-179148533 on Build GRCh38
rs950115547 Chr.3: 179148504-179148504 on Build GRCh38
rs536344415 Chr.3: 179148445-179148445 on Build GRCh38
rs983682299 Chr.3: 179148811-179148811 on Build GRCh38
rs971211185 Chr.3: 179148646-179148646 on Build GRCh38
rs199558776 Chr.3: 179148627-179148627 on Build GRCh38
rs889554970 Chr.3: 179148754-179148754 on Build GRCh38
rs937339489 Chr.3: 179148529-179148530 on Build GRCh38
rs1019591513 Chr.3: 179148484-179148484 on Build GRCh38
rs988672985 Chr.3: 179148493-179148493 on Build GRCh38
rs957327249 Chr.3: 179148619-179148619 on Build GRCh38
rs1023504355 Chr.3: 179148769-179148769 on Build GRCh38
rs905822762 Chr.3: 179148391-179148391 on Build GRCh38
rs999416097 Chr.3: 179148747-179148747 on Build GRCh38
rs111490724 Chr.3: 179148657-179148657 on Build GRCh38
rs1049079051 Chr.3: 179148675-179148675 on Build GRCh38
rs866139672 Chr.3: 179148813-179148813 on Build GRCh38
rs953708720 Chr.3: 179148446-179148446 on Build GRCh38
rs909167534 Chr.3: 179148679-179148679 on Build GRCh38
rs913045764 Chr.3: 179148495-179148495 on Build GRCh38
rs1033061125 Chr.3: 179148614-179148614 on Build GRCh38
rs961131620 Chr.3: 179148433-179148433 on Build GRCh38
rs1007026070 Chr.3: 179148767-179148768 on Build GRCh38
rs893765585 Chr.3: 179148752-179148752 on Build GRCh38
rs1051692921 Chr.3: 179148719-179148719 on Build GRCh38
rs1015018950 Chr.3: 179148426-179148427 on Build GRCh38
rs997619412 Chr.3: 179148737-179148737 on Build GRCh38
rs568092051 Chr.3: 179148533-179148533 on Build GRCh38
rs149946088 Chr.3: 179148542-179148542 on Build GRCh38
rs940961951 Chr.3: 179148723-179148723 on Build GRCh38
rs575502798 Chr.3: 179148565-179148565 on Build GRCh38
rs564749078 Chr.3: 179148553-179148553 on Build GRCh38
rs953865749 Chr.3: 179148552-179148552 on Build GRCh38
rs985818067 Chr.3: 179148451-179148451 on Build GRCh38
rs1032677804 Chr.3: 179148751-179148751 on Build GRCh38
rs1029361984 Chr.3: 179148551-179148551 on Build GRCh38

View Full Product Details