Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 68215823-68216330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R1
Entrez Gene ID
5295
Gene Name
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
UniGene
Hs.132225
Gene Chromosome Location
Chr.5: 68215737-68301821 on Build GRCh38
Transcripts
NM_181523.2 XM_005248542.3 XM_017009585.1

View Full Product Details