Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 154547506-154548067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
G6PD
Entrez Gene ID
2539
Gene Name
glucose-6-phosphate dehydrogenase
Gene Aliases
G6PD1
UniGene
Hs.461047
Gene Chromosome Location
Chr.X: 154531390-154547586 on Build GRCh38
Transcripts
NM_001042351.2 NM_001099856.4 NM_001099857.2 NM_001145255.2 NM_001321396.1 NM_001321397.1 NM_003639.4

Gene Symbol
IKBKG
Entrez Gene ID
8517
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase gamma
Gene Aliases
AMCBX1, FIP-3, FIP3, Fip3p, IKK-gamma, IKKAP1, IKKG, IMD33, IP, IP1, IP2, IPD2, NEMO, ZC2HC9
UniGene
Hs.43505
Gene Chromosome Location
Chr.X: 154542240-154565046 on Build GRCh38
Transcripts
NM_001042351.2 NM_001099856.4 NM_001099857.2 NM_001145255.2 NM_001321396.1 NM_001321397.1 NM_003639.4

SNP Detail

SNP ID Chromosome Location
rs956479198 Chr.X: 154547991-154547991 on Build GRCh38
rs996879604 Chr.X: 154547577-154547577 on Build GRCh38
rs1028298663 Chr.X: 154547592-154547592 on Build GRCh38
rs5986992 Chr.X: 154547892-154547892 on Build GRCh38
rs782179421 Chr.X: 154547870-154547870 on Build GRCh38
rs782776735 Chr.X: 154548010-154548010 on Build GRCh38
rs1038592077 Chr.X: 154547911-154547911 on Build GRCh38
rs781892396 Chr.X: 154547671-154547671 on Build GRCh38
rs938965249 Chr.X: 154547772-154547772 on Build GRCh38
rs56262786 Chr.X: 154547940-154547940 on Build GRCh38
rs782640672 Chr.X: 154547903-154547903 on Build GRCh38
rs782435077 Chr.X: 154547566-154547566 on Build GRCh38
rs56191948 Chr.X: 154547875-154547875 on Build GRCh38
rs980494975 Chr.X: 154547727-154547727 on Build GRCh38
rs1049764825 Chr.X: 154547943-154547943 on Build GRCh38
rs782526997 Chr.X: 154547878-154547878 on Build GRCh38
rs111827785 Chr.X: 154547570-154547570 on Build GRCh38
rs782425753 Chr.X: 154547844-154547844 on Build GRCh38
rs990718180 Chr.X: 154547683-154547683 on Build GRCh38
rs73641104 Chr.X: 154547962-154547962 on Build GRCh38
rs898304404 Chr.X: 154547919-154547919 on Build GRCh38
rs183785399 Chr.X: 154548043-154548043 on Build GRCh38
rs956269153 Chr.X: 154547677-154547677 on Build GRCh38
rs898562211 Chr.X: 154547572-154547572 on Build GRCh38
rs908451546 Chr.X: 154547884-154547884 on Build GRCh38
rs915027800 Chr.X: 154547703-154547703 on Build GRCh38
rs782037926 Chr.X: 154547869-154547869 on Build GRCh38
rs955344672 Chr.X: 154547648-154547648 on Build GRCh38
rs782492736 Chr.X: 154547588-154547588 on Build GRCh38
rs968184978 Chr.X: 154547721-154547721 on Build GRCh38
rs782642855 Chr.X: 154547724-154547724 on Build GRCh38
rs891126471 Chr.X: 154547944-154547944 on Build GRCh38
rs1031842193 Chr.X: 154547668-154547668 on Build GRCh38
rs1008178831 Chr.X: 154547961-154547961 on Build GRCh38
rs1004414513 Chr.X: 154547536-154547536 on Build GRCh38
rs1008440659 Chr.X: 154547650-154547650 on Build GRCh38

View Full Product Details