Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138654496-138654995 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3CB
Entrez Gene ID
5291
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
Gene Aliases
P110BETA, PI3K, PI3KBETA, PIK3C1
UniGene
Hs.239818
Gene Chromosome Location
Chr.3: 138652698-138834938 on Build GRCh38
Transcripts
NM_001256045.1 NM_006219.2 XM_005247530.2 XM_006713659.3 XM_011512895.2 XM_011512896.1 XM_017006619.1 XM_017006620.1 XM_017006621.1

SNP Detail

SNP ID Chromosome Location
rs925254061 Chr.3: 138654797-138654797 on Build GRCh38
rs573313050 Chr.3: 138654629-138654629 on Build GRCh38
rs915879465 Chr.3: 138654902-138654906 on Build GRCh38
rs964434070 Chr.3: 138654559-138654559 on Build GRCh38
rs927004062 Chr.3: 138654963-138654963 on Build GRCh38
rs950576651 Chr.3: 138654872-138654872 on Build GRCh38
rs1030595059 Chr.3: 138654689-138654689 on Build GRCh38
rs867389960 Chr.3: 138654837-138654837 on Build GRCh38
rs938963936 Chr.3: 138654577-138654577 on Build GRCh38
rs1030274046 Chr.3: 138654871-138654871 on Build GRCh38
rs892021988 Chr.3: 138654608-138654608 on Build GRCh38
rs1006974645 Chr.3: 138654735-138654735 on Build GRCh38
rs983125845 Chr.3: 138654885-138654885 on Build GRCh38
rs948796406 Chr.3: 138654906-138654906 on Build GRCh38
rs990150741 Chr.3: 138654821-138654821 on Build GRCh38
rs35498086 Chr.3: 138654808-138654809 on Build GRCh38
rs182236626 Chr.3: 138654604-138654604 on Build GRCh38
rs1010195036 Chr.3: 138654626-138654626 on Build GRCh38
rs778245232 Chr.3: 138654760-138654760 on Build GRCh38
rs963479095 Chr.3: 138654918-138654918 on Build GRCh38
rs564515338 Chr.3: 138654804-138654804 on Build GRCh38
rs536058625 Chr.3: 138654726-138654726 on Build GRCh38
rs938868922 Chr.3: 138654966-138654966 on Build GRCh38
rs978311360 Chr.3: 138654768-138654768 on Build GRCh38
rs760416244 Chr.3: 138654536-138654536 on Build GRCh38
rs997190600 Chr.3: 138654567-138654567 on Build GRCh38
rs1015924254 Chr.3: 138654897-138654897 on Build GRCh38
rs766146066 Chr.3: 138654803-138654803 on Build GRCh38
rs188032238 Chr.3: 138654767-138654767 on Build GRCh38
rs997321990 Chr.3: 138654816-138654816 on Build GRCh38
rs544603773 Chr.3: 138654774-138654774 on Build GRCh38
rs979860742 Chr.3: 138654920-138654920 on Build GRCh38
rs1043027782 Chr.3: 138654647-138654647 on Build GRCh38
rs140690084 Chr.3: 138654748-138654748 on Build GRCh38
rs1017223418 Chr.3: 138654547-138654547 on Build GRCh38
rs904638945 Chr.3: 138654661-138654661 on Build GRCh38

View Full Product Details