Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 138663707-138664286 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3CB
Entrez Gene ID
5291
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit beta
Gene Aliases
P110BETA, PI3K, PI3KBETA, PIK3C1
UniGene
Hs.239818
Gene Chromosome Location
Chr.3: 138652698-138834938 on Build GRCh38
Transcripts
NM_001256045.1 NM_006219.2 XM_005247530.2 XM_006713659.3 XM_011512895.2 XM_011512896.1 XM_017006619.1 XM_017006620.1 XM_017006621.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6935246 c.2674G>T p.D892Y Chr.3: 138664028-138664028 on Build GRCh38
1180094 c.2750T>G p.I917S Chr.3: 138663952-138663952 on Build GRCh38
6562416 c.2771_2772insA p.N924fs*14 Chr.3: 138663930-138663931 on Build GRCh38
7311702 c.2786A>G p.K929R Chr.3: 138663916-138663916 on Build GRCh38
7447218 c.2788_2789insA p.T930fs*8 Chr.3: 138663913-138663914 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146109749 Chr.3: 138663921-138663921 on Build GRCh38
rs549635648 Chr.3: 138663855-138663855 on Build GRCh38
rs900054430 Chr.3: 138663730-138663730 on Build GRCh38
rs370214055 Chr.3: 138664078-138664078 on Build GRCh38
rs954941443 Chr.3: 138663762-138663762 on Build GRCh38
rs780323598 Chr.3: 138663857-138663860 on Build GRCh38
rs980533025 Chr.3: 138663827-138663827 on Build GRCh38
rs28763909 Chr.3: 138664171-138664171 on Build GRCh38
rs761165974 Chr.3: 138664023-138664023 on Build GRCh38
rs567273050 Chr.3: 138663772-138663772 on Build GRCh38
rs373689098 Chr.3: 138663859-138663859 on Build GRCh38
rs927788195 Chr.3: 138664262-138664262 on Build GRCh38
rs142933486 Chr.3: 138664015-138664015 on Build GRCh38
rs1004309772 Chr.3: 138663832-138663832 on Build GRCh38
rs899207960 Chr.3: 138663890-138663890 on Build GRCh38
rs1057086846 Chr.3: 138663846-138663846 on Build GRCh38
rs751768585 Chr.3: 138663884-138663884 on Build GRCh38
rs774423662 Chr.3: 138664007-138664007 on Build GRCh38
rs189066091 Chr.3: 138663773-138663773 on Build GRCh38
rs1031235812 Chr.3: 138664105-138664105 on Build GRCh38
rs891133224 Chr.3: 138664178-138664178 on Build GRCh38
rs144592605 Chr.3: 138664263-138664263 on Build GRCh38
rs777541291 Chr.3: 138664008-138664008 on Build GRCh38
rs924331971 Chr.3: 138664127-138664127 on Build GRCh38
rs764552777 Chr.3: 138664071-138664071 on Build GRCh38
rs949717088 Chr.3: 138664026-138664026 on Build GRCh38
rs955678201 Chr.3: 138663808-138663808 on Build GRCh38
rs996400252 Chr.3: 138663898-138663898 on Build GRCh38
rs200500832 Chr.3: 138663892-138663892 on Build GRCh38
rs575833393 Chr.3: 138664222-138664222 on Build GRCh38
rs148771341 Chr.3: 138663981-138663981 on Build GRCh38
rs1008604539 Chr.3: 138663767-138663767 on Build GRCh38
rs932912916 Chr.3: 138664168-138664168 on Build GRCh38
rs961872349 Chr.3: 138663857-138663857 on Build GRCh38
rs192174292 Chr.3: 138664154-138664154 on Build GRCh38
rs867451277 Chr.3: 138664233-138664233 on Build GRCh38
rs372371923 Chr.3: 138664244-138664244 on Build GRCh38
rs375624065 Chr.3: 138663888-138663888 on Build GRCh38
rs759710753 Chr.3: 138664009-138664009 on Build GRCh38
rs1004597816 Chr.3: 138664181-138664181 on Build GRCh38
rs755135257 Chr.3: 138664015-138664016 on Build GRCh38
rs778504072 Chr.3: 138663799-138663799 on Build GRCh38
rs989264626 Chr.3: 138663781-138663781 on Build GRCh38
rs747984528 Chr.3: 138663897-138663897 on Build GRCh38
rs182805991 Chr.3: 138664184-138664184 on Build GRCh38
rs769545721 Chr.3: 138663899-138663899 on Build GRCh38
rs771013500 Chr.3: 138663986-138663986 on Build GRCh38
rs956685543 Chr.3: 138663779-138663779 on Build GRCh38
rs980605110 Chr.3: 138664238-138664238 on Build GRCh38
rs973183095 Chr.3: 138663880-138663880 on Build GRCh38
rs751173883 Chr.3: 138663863-138663863 on Build GRCh38
rs991602361 Chr.3: 138663988-138663988 on Build GRCh38
rs571577960 Chr.3: 138664079-138664079 on Build GRCh38
rs958906406 Chr.3: 138664223-138664223 on Build GRCh38
rs1015593330 Chr.3: 138663851-138663851 on Build GRCh38
rs775468022 Chr.3: 138664150-138664150 on Build GRCh38
rs997281441 Chr.3: 138663756-138663756 on Build GRCh38
rs770301071 Chr.3: 138664061-138664061 on Build GRCh38
rs777754324 Chr.3: 138663954-138663954 on Build GRCh38
rs1054612826 Chr.3: 138664151-138664151 on Build GRCh38
rs558493712 Chr.3: 138664063-138664063 on Build GRCh38
rs991429142 Chr.3: 138664247-138664247 on Build GRCh38
rs555983498 Chr.3: 138664221-138664221 on Build GRCh38
rs939263133 Chr.3: 138663837-138663837 on Build GRCh38
rs746296681 Chr.3: 138664216-138664216 on Build GRCh38
rs533960524 Chr.3: 138664149-138664149 on Build GRCh38
rs898573159 Chr.3: 138664191-138664191 on Build GRCh38

View Full Product Details