Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 68273329-68273503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIK3R1
Entrez Gene ID
5295
Gene Name
phosphoinositide-3-kinase regulatory subunit 1
Gene Aliases
AGM7, GRB1, IMD36, p85, p85-ALPHA
UniGene
Hs.132225
Gene Chromosome Location
Chr.5: 68215737-68301821 on Build GRCh38
Transcripts
NM_181523.2 XM_005248542.3 XM_011543493.2 XM_017009585.1 XM_017009586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5945951 c.410A>G p.E137G Chr.5: 68273465-68273465 on Build GRCh38
6229516 c.351T>G p.D117E Chr.5: 68273406-68273406 on Build GRCh38
6937716 c.360G>T p.E120D Chr.5: 68273415-68273415 on Build GRCh38
3683873 c.366T>C p.F122F Chr.5: 68273421-68273421 on Build GRCh38
3683872 c.362A>T p.Q121L Chr.5: 68273417-68273417 on Build GRCh38
View More

View Full Product Details